Zebrafish E2 (Estradiol) ELISA Kit

Supplier: AFG Bioscience

EK248153
77526-722EA 609.41 CAD
77526-722
Zebrafish E2 (Estradiol) ELISA Kit
Assays ELISAs
New Product

Order Now

SPECIFICATIONS

Target protein E2 (Estradiol)
Environmentally Preferable
Primary antibody reactivity
Assay Type Competitive
Host Zebrafish
Sensitivity 4.45 pg/ml
Detection range 15.63 - 1000 pg/ml
Sample type Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
Time to Results 2 h
Regulatory status RUO
Storage temperature 2…8 °C
Shelf life 6 months (4 °C); 12 months (–20 °C)
Application Endocrinology, Reproductive science, Hormone metabolism

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR