Human YWHAq (Tyrosine 3/Tryptophan 5 Monooxygenase Activation Protein Theta) ELISA Kit

Supplier: AFG Bioscience

EK246745
77523-906EA 609.41 CAD
77523-906
Human YWHAq (Tyrosine 3/Tryptophan 5 Monooxygenase Activation Protein Theta) ELISA Kit
Assays ELISAs

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein YWHAq (Tyrosine 3/Tryptophan 5 Monooxygenase Activation Protein Theta)
Primary antibody reactivity
Assay Type Sandwich
Host Human
Sensitivity 0.053 ng/ml
Detection range 0.16 - 10 ng/ml
Sample type Serum, plasma, tissue homogenates and other biological fluids
Time to Results 3.5 h
Regulatory status RUO
Storage temperature 2…8 °C
Shelf life 6 months (4 °C); 12 months (–20 °C)
Application Enzyme and Kinase

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR