Mouse EIF2aK3 (Eukaryotic Translation Initiation Factor 2 Alpha Kinase 3) ELISA Kit

Supplier: AFG Bioscience

EK246468
77523-352EA 609.41 CAD
77523-352
Mouse EIF2aK3 (Eukaryotic Translation Initiation Factor 2 Alpha Kinase 3) ELISA Kit
Assays ELISAs

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein EIF2aK3 (Eukaryotic Translation Initiation Factor 2 Alpha Kinase 3)
Primary antibody reactivity
Assay Type Sandwich
Host Mouse
Sensitivity 0.108 ng/ml
Detection range 0.32 - 20 ng/ml
Sample type Tissue homogenates and other biological fluids
Time to Results 3.5 h
Regulatory status RUO
Storage temperature 2…8 °C
Shelf life 6 months (4 °C); 12 months (–20 °C)
Application Developmental science

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR