Human ARHGDIa (Rho GDP Dissociation Inhibitor Alpha) ELISA Kit

Supplier: AFG BIOSCIENCE LLC

EK242670
77515-756EA 609.41 CAD
77515-756
Human ARHGDIa (Rho GDP Dissociation Inhibitor Alpha) ELISA Kit
Assays ELISAs
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein ARHGDIa (Rho GDP Dissociation Inhibitor Alpha)
Primary antibody reactivity
Assay Type Sandwich
Host Human
Sensitivity 0.066 ng/ml
Detection range 0.16 - 10 ng/ml
Sample type Tissue homogenates and other biological fluids
Time to Results 3.5 h
Regulatory status RUO
Storage temperature 2…8 °C
Shelf life 6 months (4 °C); 12 months (–20 °C)
Application Tumor immunity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR