Rat IL3 (Interleukin 3) ELISA Kit

Supplier: AFG Bioscience

EK242058
77514-532EA 609.41 CAD
77514-532
Rat IL3 (Interleukin 3) ELISA Kit
Assays ELISAs

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein IL3 (Interleukin 3)
Primary antibody reactivity
Assay Type Sandwich
Host Rat
Sensitivity 2.8 pg/ml
Detection range 7.82 - 500 pg/ml
Sample type Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
Time to Results 3.5 h
Regulatory status RUO
Storage temperature 2…8 °C
Shelf life 6 months (4 °C); 12 months (–20 °C)
Application Cytokine, Infection immunity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR