Syringes for Automatic Liquid Handling, XLP
Supplier: INNOVATIVE LABOR SYSTEME GMBH
2679011 2679015 2679031 2679035 2679051 2679055 2679071 2679075 2679076
77506-518EA 130.22 CAD
77506-518 77506-520 77506-524 77506-526 77506-528 77506-530 77506-534 77506-536 77506-538
Syringes for Automatic Liquid Handling, XLP
Syringes Syringes for Chromatography
These syringes comes with the three types of seals.
  • Front fitting: ¹/₄"-28 UNF
  • Glass barrel: Borosilicate 3.3
  • Stroke: 60 mm
PTFE-seal: Chemically resistant; heavy duty syringes.
PTFE-seal with silicone: Specially designed for handling of strong buffers and other aqueous solutions. The modified PTFE-seals guaranteed, extended lifetime under heavy duty conditions.
HDPE-seal: Specially designed for handling of strong buffers and other aqueous solutions. The special PE-seals run smoother than conventional seals. Good resistance to buffers andMore Product Information