Syringes for Automatic Liquid Handling, H-KP
Supplier: INNOVATIVE LABOR SYSTEME GMBH
2608815 2608816 2608915 2608076 2608091 2608096 2608105 2608125 2608515 2608516 2608615 2608616 2608715 2608015 2608015-HT 2608035 2608036 2608040 2608055 2608056 2608075
77506-310EA 174.23 CAD
77506-310 77506-312 77506-314 77506-290 77506-292 77506-294 77506-296 77506-298 77506-300 77506-302 77506-304 77506-306 77506-308 77506-276 77506-636 77506-278 77506-280 77506-282 77506-284 77506-286 77506-288
Syringes for Automatic Liquid Handling, H-KP
Syringes Syringes for Chromatography
Series H-KP automatic liquid handling syringe with a glass barrel made from borosilicate 3.3 designed for volumes of 25 µl to 50 ml. 60 mm stroke and the syringe has a PTFE (polytetrafluoroethylene) plunger tip. A high chemical resistance makes this syringe a perfect heavy-duty syringes.
The syringe is manufactured from high-quality DURAN 3.3 borosilicate glass, that is heat and shock resistant, which ultimately protects against breakage from shock and sudden or extreme fluctuations in temperature. This type of ultra-low expansion glass endow the syringe with maximum chemical resistance,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable