iFluor® 670 maleimide
Supplier: AAT BIOQUEST INC
72502
77463-104EA 371.92 CAD
77463-104
iFluor® 670 maleimide
iFluor® 670 maleimide
iFluor® 670 dyes have fluorescence excitation and emission maxima of ~670 nm and ~690 nm respectively.
  • Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
Caution: For Research Use Only
Order Now
SPECIFICATIONS
Environmentally Preferable