FAM SSL-NHS ester, ReadiCleave™
Supplier: AAT BIOQUEST INC
7041
77463-040EA 674.53 CAD
77463-040
FAM SSL-NHS ester, ReadiCleave™
FAM SSL-NHS ester
ReadiCleave™ FAM SSL-NHS ester contains a S-S linker that can be cleaved with TCEP or DTT to remove the fluorescein (FITC) fluorophore from the target molecule.
  • Stable for 12 months
AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.
Caution: For Research Use Only