iFluor® 405 styramide
Supplier: AAT BIOQUEST INC
44908
77462-270EA 517.01 CAD
77462-270
iFluor® 405 styramide
iFluor® 405 styramide
iFluor® 405 Styramide is a superior replacement for Alexa Fluor 405 tyramide or other spectrally similar fluorescent tyramide conjugates or TSA reagents.
  • Stable for 12 months
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide (TSA) reagents.
Caution: For Research Use Only