5.1 CPG [TQ5.1 CPG], Tide Quencher™ (500 A)
Supplier: AAT BIOQUEST INC
2084
77461-776EA 331.83 CAD
77461-776
5.1 CPG [TQ5.1 CPG], Tide Quencher™ (500 A)
5.1 CPG [TQ5.1 CPG]
Tide Quencher™ 5.1 (TQ5.1) is a non-fluorescent molecule designed to efficiently quench the fluorescence of common NIR fluorophores such as Cy5, Cy5.5, Alexa Fluor® 647, Alexa Fluor® 647, iFluor® 647 and iFluor® 680.
  • Stable for 12 months
  • Optimized for FRET applications
Quality tested in-house
Caution: For Research Use Only