XFD700 acid
Supplier: AAT BIOQUEST INC
1799
77461-698EA 745.48 CAD
77461-698
XFD700 acid
XFD700 acid
XFD700 is the same molecule as the Alexa Fluor® 700 (Alexa Fluor® is the trademark of ThermoFisher).
  • Stable for 12 months
Quality tested in-house