DCS2, Nuclear Blue™ [Equivalent to SYTOX™ Blue Dead Cell Stain]
Supplier: AAT BIOQUEST INC
17546 17547
77461-620EA 325.04 CAD
77461-620 77461-622
DCS2, Nuclear Blue™ [Equivalent to SYTOX™ Blue Dead Cell Stain]
DCS2
Nuclear Blue™ DCS2 is the same molecule to SYTOX™ Blue Dead Cell Stain (SYTOX™ is a trademark of ThermoFisher).
  • Stable for 12 months
Quality tested in-house
Caution: For Research Use Only