mFluor™ Violet 450 azide
Supplier: AAT BIOQUEST INC
1690
77461-508EA 517.01 CAD
77461-508
mFluor™ Violet 450 azide
mFluor™ Violet 450 azide
mFluor™ Violet 450 dyes have fluorescence excitation and emission maxima of ~405 nm and ~450 nm respectively.
  • Stable for 12 months
AAT Bioquest's mFluor™ dyes are developed for multicolor flow cytometry-focused applications.
Caution: For Research Use Only