iFluor® 770 maleimide
Supplier: AAT BIOQUEST INC
1370
77461-458EA 558.7 CAD
77461-458
iFluor® 770 maleimide
iFluor® 770 maleimide
iFluor® 770 dyes have fluorescence excitation and emission maxima of ~777 nm and ~797 nm respectively.
  • Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, particularly antibodies.
Caution: For Research Use Only