Anti-CD4 Mouse Monoclonal Antibody [clone: SK3] (APC-Astral™813)

Supplier: Biotium

P001-A813-500
77449-102EA 690.62 CAD
77449-102
Anti-CD4 Mouse Monoclonal Antibody [clone: SK3] (APC-Astral™813)
Antibodies
CD4 (clone SK3) is a validated mouse monoclonal antibody that recognizes CD4, a membrane glycoprotein of T lymphocytes. The antibody have been validated and optimized in-house for optimal performance.

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: SK3
Conjugation: APC-Astral™813
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR