DYNEO™ DD Refrigerated/Heating Circulators
Supplier: JML SCIENCES INC CA
9021701.02 9021701.33 9021701.02.D 9021701.33.D 9021701.02.A 9021701.33.A 9021703.02 9021703.13 9021703.02.D 9021703.13.D 9021703.02.A 9021703.13.A 9021704.02 9021704.33 9021704.02.D 9021704.33.D 9021704.02.A 9021704.33.A 9021707.02.D 9021707.33.D 9021707.02.A 9021707.33.A 9021707.02 9021707.33 9021701.02 9021701.33 9021701.02.D 9021701.33.D 9021701.02.A 9021701.33.A 9021703.02 9021703.13 9021703.02.D 9021703.13.D 9021703.02.A 9021703.13.A 9021704.02 9021704.33 9021704.02.D 9021704.33.D 9021704.02.A 9021704.33.A 9021707.02.D 9021707.33.D 9021707.02.A 9021707.33.A 9021707.02 9021707.33 9021713.N1.02 9021713.N1.02.A 9021713.N1.02.D 9021713.N1.33 9021713.N1.33.A 9021713.N1.33.D 9021713.S1.02 9021713.S1.02.A 9021713.S1.02.D 9021713.S1.33 9021713.S1.33.A 9021713.S1.33.D 9021714.N1.02 9021714.N1.02.A 9021714.N1.02.D 9021714.N1.33 9021714.N1.33.A 9021714.N1.33.D 9021714.S1.02 9021714.S1.02.A 9021714.S1.02.D 9021714.S1.33 9021714.S1.33.A 9021714.S1.33.D 9021701.N1.33.A 9021701.N1.33.D 9021715.N1.02 9021715.N1.02.A 9021715.N1.02.D 9021715.N1.33 9021715.N1.33.A 9021715.N1.33.D 9021717.N1.02 9021717.N1.02.A 9021717.N1.02.D 9021717.N1.33 9021717.N1.33.A 9021717.N1.33.D 9021728.N1.02 9021728.N1.02.A 9021728.N1.02.D 9021728.N1.33 9021728.N1.33.A 9021728.N1.33.D 9021701.N1.02 9021701.N1.02.A 9021701.N1.02.D 9021701.N1.33
76289-282EA 15281.67 CAD
76289-282 76289-284 76289-286 76289-288 76289-290 76289-292 76289-294 76289-296 76289-298 76289-300 76289-302 76289-304 76289-306 76289-308 76289-310 76289-312 76289-314 76289-316 76289-318 76289-320 76289-322 76289-324 76289-326 76289-328 76289-330 76289-332 76289-334 76289-336 76289-338 76289-340 76289-342 76289-344 76289-346 76289-348 76289-350 76289-352 76289-354 76289-356 76289-358 76289-374 76289-376 76289-378 76289-380 76289-382 76289-384 CA76289-282 CA76289-284 CA76289-286 CA76289-288 CA76289-290 CA76289-292 CA76289-294 CA76289-296 CA76289-298 CA76289-300 CA76289-302 CA76289-304 CA76289-306 CA76289-308 CA76289-310 CA76289-312 CA76289-314 CA76289-316 CA76289-318 CA76289-320 CA76289-322 CA76289-324 CA76289-326 CA76289-328 CA76289-330 CA76289-332 CA76289-334 CA76289-336 CA76289-338 CA76289-340 CA76289-342 CA76289-344 CA76289-346 CA76289-348 CA76289-350 CA76289-352 CA76289-354 CA76289-356 CA76289-358 CA76289-374 CA76289-376 CA76289-378 CA76289-380 CA76289-382 CA76289-384 76558-242 76558-184 76558-172 76558-250 76558-252 76558-264 76558-230 76558-162 76558-232 76558-260 76558-258 76558-274 76558-214 76558-182 76558-256 76558-266 76558-170 76558-164 76558-176 76558-158 76558-192 76558-224 76558-202 76558-244 76558-208 76558-178 76558-226 76558-248 76558-204 76558-186 77442-100 77442-102 77442-104 77442-106 77442-108 77442-110 77442-112 77442-114 77442-116 77442-118 77442-120 77442-122 77442-124 77442-126 77442-128 77442-130 77442-132 77442-134 77442-136 77442-138 77442-092 77442-094 77442-096 77442-098
DYNEO™ DD Refrigerated/Heating Circulators
Circulating Thermostats Closed Circulator Systems Temperature Control Systems
DYNEO™ DD refrigerated/heating circulator combinations with stainless steel bath tanks for internal or external temperature applications from –20 to 200 to –50 to 200 °C.
  • Infinitely adjustable pressure pump
  • Easy switching between internal and external circulation
  • Large color TFT display, multilingual interface
  • Integrated programmer
  • PT100 port facilitates external temperature control via PT100 sensor
  • USB port
  • Optional RS232 interface or analog interfaces
For internal and external temperature applications. High quality stainless steel bath tanks withMore Product Information