Accessories for Compresstome® Tissue Glue
Supplier: PRECISIONARY INSTRUMENTS LLC
VFVMGLUECANAL
77435-524EA 20.35 CAD
77435-524
Accessories for Compresstome® Tissue Glue
Microtomes and Microtome Assemblies Microtomes
Glue is designed to secure the samples onto the specimen tube prior to embedding the sample in agarose.
  • One 0.07 oz. tube with drop applicator
Instant Krazy Glue (cyanoacrylate) recommended for tissue specimens for use with Compresstome tissue slicers.