Compresstome® VF-800-0Z Vibratome
Supplier: PRECISIONARY INSTRUMENTS LLC
VF8000ZVMNC
77435-380EA 156807.07 CAD
77435-380
Compresstome® VF-800-0Z Vibratome
Microtomes and Microtome Assemblies Microtomes
The Compresstome VF-800-0Z is a vibrating microtome for cutting tissue slices with Auto Zero-Z technology.
  • Fully automated for high through-put tissue sectioning
  • Up to 100 mm tissue diameter
  • Full one year warranty
This is the large diameter Compresstome model for large sample slicing and high throughput slicing. Includes VF-800-0Z vibrating microtome and one blade holder.