Accessories for Rotary Microtome Handwheels
Supplier: PRECISIONARY INSTRUMENTS LLC
RF600COARSEWHL RF600HANDWHEEL RF800LARGEWHL RF1000HNDWHEEL
77435-322EA 281.59 CAD
77435-322 77435-324 77435-354 77435-296
Accessories for Rotary Microtome Handwheels
Microtomes and Microtome Assemblies Microtomes
An assortment of rotary microtome handwheels for use in manual slicing operations.
The Rotary Microtome RF-600 coarse handwheel is used to move the specimen head in fast forward and fast reverse directions. This unit allows for fast advancement of the specimen head clamp that holds the sample.
The Rotary Microtome RF-600, RF-800, and RF-1000 large handwheels are used to move the specimen head up and down for cutting.
Ordering information: Large handwheel units are supplied with handwheel and handwheel handle.
Order Now
SPECIFICATIONS
Environmentally Preferable