Navigator® Portable Balances
Supplier: Ohaus
30456410 30456411 30456412 30456413 30456414 30456415 30456416 30456417 30456418 30456419 30456420 30456421 30467960 30467961 30642238 30642239 30642240 30642241 30642242 30642243 30642244
76377-610EA 672.45 CAD
76377-610 76377-612 76377-614 76377-616 76377-618 76377-620 76377-622 76377-624 76377-626 76377-628 76377-630 76377-632 77434-972 77434-974 77435-118 77435-120 77435-122 77435-124 77435-126 77435-128 77435-130
Navigator® Portable Balances
Balances and Scales
Multi-purpose portable balances suitable for everyday weighing. Navigator offers essential weighing applications at a competitive price point.
  • Advanced weighing technology offers one-second stabilization for fast and accurate weighing results, improving operator efficiency, productivity and throughput
  • Overload protection system that can withstand loads up to four times the rated capacity safeguards the balance
OHAUS raises the bar in price-performance ratio with the Navigator series which is perfect for general weighing applications in laboratory,More Product Information