Defender™ 6000 Industrial Washdown Bench Scales
Supplier: Ohaus
30631706 30631707 30631718 30631719 30575563 30575564 30575565 30575566 30575567 30631720 30631721 30631722 30631723 30631724
77435-088EA 3527.73 CAD
77435-088 77435-090 77435-092 77435-094 77435-038 77435-040 77435-042 77435-044 77435-046 77435-096 77435-098 77435-100 77435-102 77435-104
Defender™ 6000 Industrial Washdown Bench Scales
Balances and Scales
Advanced IP68/IP69k washdown bench scales. The durable Defender 6000 stands up to harsh cleaning in processing and packaging environments with IP 68 immersion/ IP69K washdown protection, 316 stainless steel base and hermetically-sealed load cell.
  • Supports HACCP systems with a 316 stainless steel base and sturdy indicator providing corrosion protection in food, chemical processing and packaging
  • Versatile scales that include fast weight display and multiple software modes
The Defender 6000 series extreme washdown bench scale is specifically designed forMore Product Information