Stainless Steel Indicator for Industial Applications, Defender 6000
Supplier: Ohaus
30575584
77435-062EA 1799.54 CAD
77435-062
Stainless Steel Indicator for Industial Applications, Defender 6000
Balances and Scales
Washdown bench stainless steel indicator for standard industrial applications. Defender 6000 indicator, constructed from 316 stainless steel, stands up to harsh cleaning in processing and packaging environments with IP 68 immersion/IP 69K washdown protection.
  • White LED displays clear results for fast processing
  • The indicator's configurable color bar matrix allows for detailed package weight checking to ensure compliance
  • Knife-proof numeric touch keypad, indicator is suitable in harsh processing environments
The Defender 6000 Series extreme washdownMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable