Defender™ 6000 Industrial Bench Scales
Supplier: Ohaus
30626682 30626683 30626684 30631703 30631704 30631705 30575562 30575568 30575569 30575570 30575571 30631730 30575572 30575573 30608733 30608734 30608735 30626679 30626680 30626681 30572969 30572970 30572971 30631725 30631726 30631727 30631728 30631729
77435-076EA 3354.39 CAD
77435-076 77435-078 77435-080 77435-082 77435-084 77435-086 77435-036 77435-048 77435-050 77435-052 77435-054 77435-116 77435-056 77435-058 77435-064 77435-066 77435-068 77435-070 77435-072 77435-074 77435-030 77435-032 77435-034 77435-106 77435-108 77435-110 77435-112 77435-114
Defender™ 6000 Industrial Bench Scales
Balances and Scales
Bench scales for standard industrial applications. Defender 6000 indicator constructed of polycarbonate stands up to harsh cleaning in processing and packaging environments with IP68 immersion/IP69K washdown protection.
  • Large, bright backlit LCD clearly displays results for fast processing
  • The color bar can be used for quick checking of package weights for fast go/no-go alert
  • Durably constructed with a 304 stainless steel platform, frame, IP67 loadcell
The Defender 6000 series Hybrid Washdown bench scale is ideal for a multitude of applications,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable