Analytical Balances, PR
Supplier: Ohaus
30430060 30430061 30430062 30529065 30529066
77434-832EA 3063.21 CAD
77434-832 77434-834 77434-836 77434-988 77434-990
Analytical Balances, PR
Balances and Scales
Designed for routine weighing applications. The PR is equipped with 3 essential weighing modes, RS-232 connectivity for data transfer and storage and internal calibration (in select models), making it ideal for routine weighing applications.
  • Easy-to-read, bright backlit display and a simple user interface
  • Incredibly easy to operate, with almost no training required
  • The small footprint saves desktop space while providing a large weighing surface
  • Durable construction and features a stainless steel pan to withstand day-to-day use in the workplace