Defender 5000 Industrial Washdown Bench Scales
Supplier: Ohaus
30467614 30467615 30467616 30467617 30467618 30467619 30467620 30467621 30461673 30461674 30461675 30461676 30461677 30461678 30461679 30461680 30461681 30461682 30461683 30461684 30461685 30461686 30461687 30461688 30461689 30461690 30461691 30461692 30470990 30461464 30461465 30461466 30461467 30461661 30461662 30461663 30461664 30461665 30461668 30461669 30461670 30461671 30461672 30461693 30461694 30461695 30461696 30461697 30461698 30461699 30461700 30461701 30461702 30461703 30461704 30461705 30461706 30467608 30467609 30467610 30467611 30467612 30467613
77434-956EA 4074.76 CAD
77434-956 77434-958 77434-960 77434-962 77434-964 77434-966 77434-968 77434-970 77434-876 77434-878 77434-880 77434-882 77434-884 77434-886 77434-888 77434-890 77434-892 77434-894 77434-896 77434-898 77434-900 77434-902 77434-904 77434-906 77434-908 77434-910 77434-912 77434-914 77434-986 77434-846 77434-848 77434-850 77434-852 77434-856 77434-858 77434-860 77434-862 77434-864 77434-866 77434-868 77434-870 77434-872 77434-874 77434-916 77434-918 77434-920 77434-922 77434-924 77434-926 77434-928 77434-930 77434-932 77434-934 77434-936 77434-938 77434-940 77434-942 77434-944 77434-946 77434-948 77434-950 77434-952 77434-954
Defender 5000 Industrial Washdown Bench Scales
Balances and Scales
Multifunctional stainless steel washdown bench scale for standard industrial applications. User-friendly models with an easy-to-use keypad, large backlit display and plain text on-screen prompts which guide you through operation.
  • Durable construction with stainless steel platform, frame and IP 68 indicator
  • IP67 stainless steel loadcell (NTEP/MC/OIML/EC Type single range) or aluminum load cell (OIML/EC Type dual range)
  • Features common industrial modes and supports a 50,000-item library with 1000 user profiles for easy data management
  • Incudes GMP/GLP with
More Product Information