ScrubberStand™ Stackable Stands
Supplier: Labconco
4618000 4618001
77400-898EA 1690.6 CAD
77400-898 77400-900
ScrubberStand™ Stackable Stands
Glassware Washers
Double your washing capacity by stacking 2 glassware washers, reduce floor space.
  • Space efficiency: Optimizes space by enabling vertical stacking of washers
  • Increased capacity: Doubles washing capacity without expanding laboratory area
  • Safe: ETL certification eliminates any worries over the possibility of tipping
  • Ease of installation: Comes already assembled
  • Compatibility: Designed to work seamlessly with our existing range of glassware washers
Maximize glassware washing capacity without using extra floor space. Stack 2 Freestanding Washers with aMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable