Cryo Tube Storage Box PC, 81-Place
Supplier: Heathrow Scientific
HS23381B
77395-386PK 74.48 CAD
77395-386
Cryo Tube Storage Box PC, 81-Place
Boxes Cryoboxes
Safely store cryogenic vials.
  • Our polycarbonate boxes are designed with unique features to help orient, identify and access vials with ease
  • Alphanumeric labeling on lid or base (depending on the unit) for quick location and retrieval of tubes
  • Liquid nitrogen vent and drainage holes
  • Magnet on top of forceps enables easy storing on metal surfaces
  • Stackable, saves valuable space in the lab
The Arctic Squares® polycarbonate cryostorage box safely stores vials from −196 to +121 °C in mechanical freezers as well as liquid nitrogen. These polycarbonate boxesMore Product Information