Reagent Reservoirs, 25 ml, Dual
Supplier: Heathrow Scientific
120651 120652
77395-276CS 1285.78 CAD
77395-276 77395-278
Reagent Reservoirs, 25 ml, Dual
Reagent Reservoirs
Reagent Reservoirs by Heathrow Scientific offer a fresh and modern design with enhanced features to streamline pipetting procedures.
  • Superior durability with enhanced walls and crush-resistant 'turrets' at the corners
  • Easy handling with tabs to prevent stacked reservoirs from sticking together
  • Graduation markings on the top edge and inner walls allow measurements of liquid to be easily identified and reduce reagent waste
  • Pour-off spouts conveniently located on all four corners for easy spill-free pouring and reduced dripping
  • Divided reservoir design allows
More Product Information