BioX Tissue Culture Flasks
Supplier: TWD Scientific
BX07-8025 BX07-8075 BX07-8182 BX07-8300
77394-954CS 341.2 CAD
77394-954 77394-956 77394-958 77394-960
BioX Tissue Culture Flasks
Flasks Culture Flasks
Ideal for the cultivation and observation of sensitive cells.
  • Tilted neck
  • PTFE hydrophobic vent cap
  • Marked volume scales on both sides
  • Frosted writing area
  • DNase/RNase, non-pyrogenic
  • Highly transparent polystyrene
The angled neck allows the user to access the entire surface area. Easy sample identification due to the molded graduation and frosted writing patch.
Order Now
SPECIFICATIONS
Environmentally Preferable