Compact Bucket Systems, Contec®
Supplier: Contec
2942U2B 2942U3B 2942U3S 2942U2BF 2942U3BF
76199-900CS 2768.87 CAD
76199-900 76199-902 76199-904 76199-906 76199-930 77394-796 77394-798 77394-800 77786-112 77786-114
Compact Bucket Systems, Contec®
Buckets
The autoclavable Compact Bucket System is an all-in-one solution to make cleaning and disinfecting easier and more efficient. The cart is designed for use with Contec® mops such as the VertiKlean® MAX™, VertiKlean® MAX™ Sealed Edge, EasyCurve®, and QuickTask™ mopping systems.
  • On-cart storage for two mop handles and frames allows convenient storage and keeps mops out of the way
  • Mop holders keep mops securely in place when changing locations
  • Improved castors and connections significantly reduces noise during use, allowing more durable attachments with less '
More Product Information