Stilla® Nio™ Digital PCR Instrument
Supplier: STILLA TECHNOLOGIES, INC.
N30027.0 N30027.1 N30027.2
77392-280EA 281585.56 CAD
77392-280 77449-124 77574-018
Stilla® Nio™ Digital PCR Instrument
Thermal Cyclers
The Nio™ digital PCR instrument line offers a simple all-in-one digital PCR workflow that supports seven colors, flexible throughput of up to 768 samples per day with three configurations, continuous loading, and easy-to-use software.
  • All-in-one digital PCR platform
  • Up to 7-color multiplexing
  • Flexible throughput with three configurations: Nio™+, Nio™, and Nio™ E
  • User-friendly design
  • Continuous loading
  • Remote access
  • AI powered droplets count
Nio™ digital PCR system is a versatile, all-in-one, fit-for-throughput digital PCR instrument with uparalleledMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Compatible with FAM, Yakima Yellow®, Atto®550, ROX, Cy®5, Atto®700/Cy®5.5, DY-521-XL
No. of channels 7 (Fluorescent channels)
W×D×H 56×65×52 cm
Weight 61 kg