Thermo Scientific™ Serological Pipettes
Supplier: Thermo Fisher Scientific
170373N 170374N 170375N 170376N 170353N 170354N 170355N 170356N 170357N 170358N 170364N 170365N 170366N 170367N 170368N 170369N 170371N 170372N
77392-150CS 183.32 CAD
77392-150 77392-152 77392-154 77392-156 77392-122 77392-124 77392-126 77392-128 77392-130 77392-132 77392-134 77392-136 77392-138 77392-140 77392-142 77392-144 77392-146 77392-148
Thermo Scientific™ Serological Pipettes
Pipettes Serological Pipettes
Bold, clear bi-directional graduations make it easy-to-read fluid volumes and dispensing amounts. Choose from a broad selection of sizes and packaging options.
  • Material - Ultra clear, 100% virgin polystyrene for maximum clarity
  • Contaminant free - RNase/DNase free, human DNA free, non-cytotoxic, non-pyrogenic
  • Sterility - E-beam irradiation to SAL of 10⁻⁶
  • Shelf life - Five years from manufacturing date
Wide range of volumes and packaging options. Color-coded pipettes and packaging for convenient volume selection. Individually packed pipettes in eitherMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable