VWR® 16 Sample Purification System
Supplier: VWR International
77321-868 77321-870 77321-872 77321-874 77321-876 77321-878
77321-868CS 221.83 CAD
77321-868 77321-870 77321-872 77321-874 77321-876 77321-878
VWR® 16 Sample Purification System
Automated Nucleic Acid Purification Systems
VWR® 16 Sample purification system is a high throughput, automated system that improves the efficiency and reproducibility of magnetic bead-based purification protocols.
  • Compatible with all popular magnetic bead kits
  • Hands-off processing and consistent results
  • Compact design
  • Processing capacity up to 16 samples
  • Programmable with PC software or Android App
Capable of processing up to 16 samples at one time, the VWR® 16 sample purification system is easy to set up, program, and operate. Mixing, bead transfer, wash steps, and elution steps are performedMore Product Information