Defender™ 5000 Industrial Hybrid Floor Scales
Supplier: Ohaus
30837696 30837698 30837699 30837706 30837707 30837708 30716491 30716493 30716494 30716497 30716498
77321-584EA 3944.26 CAD
77321-584 77321-588 77321-590 77321-596 77321-598 77321-600 77321-470 77321-474 77321-476 77321-482 77321-484
Defender™ 5000 Industrial Hybrid Floor Scales
Balances and Scales
Defender™ 5000 Multifunctional Industrial Floor Scales. Painted carbon steel, treaded decks with four alloy steel load cells, fixed top plate 5 mm (0.196") thick with structural bracing for stiff construction.
  • Equipped with a new TD52 series indicator featuring an easy-to-use keypad, large backlit display, and plain text on-screen prompts to guide user through operation