Defender™ 3000 Industrial Washdown Floor Scales
Supplier: Ohaus
30837697 30837700 30837701 30837602 30837605 30837606
77321-586EA 8876.58 CAD
77321-586 77321-592 77321-594 77321-564 77321-570 77321-572
Defender™ 3000 Industrial Washdown Floor Scales
Balances and Scales
Defender™ 3000 Washdown Floor Scales. 304 stainless steel, flat surface decks with 4 IP68 load cells, fixed top plate 5 mm (0.196") thick with structural bracing for stiff construction.
  • Results are clearly shown on the large, bright backlit LCD display
  • In Checkweighing mode, display changes color (yellow or green or red) to alert users when within the target weight range
Order Now
SPECIFICATIONS
Environmentally Preferable