Industrial Floor Scales, Platform Only
Supplier: Ohaus
30716492 30716495 30716496 30716499 30716500
77321-472EA 6240.89 CAD
77321-472 77321-478 77321-480 77321-486 77321-488
Industrial Floor Scales, Platform Only
Balances and Scales
Defender™ 3000 washdown floor platform only. 304 stainless steel, flat surface decks with 4 IP 68 load cells, fixed top plate 5 mm (0.196 ") thick with structural bracing for stiff construction.
  • Equipped with a 20 ft EasyConnect™ cable which allows instant configuration with Defender indicators for quicker installation