Defender 3000™ Industrial Washdown Bench Scales
Supplier: Ohaus
30685209 30685210 30685211 30685197 30685198 30685180 30685181 30685216 30685217 30685222 30685223 30685182 30685191 30685192
77321-440EA 2990.83 CAD
77321-440 77321-442 77321-444 77321-416 77321-418 77321-390 77321-392 77321-454 77321-456 77321-466 77321-468 77321-394 77321-404 77321-406
Defender 3000™ Industrial Washdown Bench Scales
Balances and Scales
The OHAUS Defender™ 3000 Washdown - i-D33 bench scales are ideal for general weighing, simple counting and checkweighing applications in damp environments or when washdown cleaning is required.
  • Simple, yet rugged, stainless steel design
  • IP66 stainless steel indicator with tactile keys
  • Multicolor backlit LCD display
  • AC or standard rechargeable battery operation
  • Flexible mounting capabilities
Results are clearly shown using the large, bright backlit LCD display. In Checkweighing mode, the display changes color (yellow or green or red) to alert when itMore Product Information