Epredia™ 615 Laser Bionet Cassettes with Separate Lids
Supplier: Epredia
615-06-10LMI 615-07-10LMI 615-08-10LMI 615-09-10LMI 615-10-10LMI 615-11-10LMI 615-12-10LMI
77308-642CS 841.5 CAD
77308-642 77308-644 77308-646 77308-648 77308-650 77308-652 77308-654
Epredia™ 615 Laser Bionet Cassettes with Separate Lids
Embedding Cassettes Cassettes for Labeling Instruments
Manufactured specifically for laser printing, the series 515 Bionet biopsy cassettes come with separate lids and are available in 12 colors.
  • Series 615 laser cassettes are designed for use on both the Epredia™ NOVA™ and VEGA™ laser printers
  • They contain a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Comes in 12 different colors: white, yellow, blue, pink, green, orange, gray, aqua, lilac, beige, ochre and red with color-matched lids
  • Taped stacks of 40 cassettes; 25 stacks per case for a total of 1000 cassettes and
More Product Information