PROCLEAR Polypropylene DEMI Capsules
Supplier: PARKER HANNIFIN TE
PEPPE-01GG-PS3-FB PEPPE-05GG-PS3-FB PEPPA-01GG-PN3 PEPPA-01GG-PS3 PEPPA-01HH-PN3 PEPPA-01HH-PS3 PEPPA-03GG-PN3 PEPPA-03GG-PS3 PEPPA-03HH-PN3 PEPPA-03HH-PS3 PEPPA-05GG-PN3 PEPPA-05GG-PS3 PEPPE-01GG-PN3 PEPPE-01GG-PS3 PEPPE-01HH-PN3 PEPPE-01HH-PS3 PEPPE-03GG-PN3 PEPPE-03GG-PS3 PEPPE-03HH-PN3 PEPPE-03HH-PS3 PEPPE-05GG-PN3 PEPPE-05GG-PS3 PEPPB-01GG-PN3 PEPPB-01GG-PS3 PEPPB-01HH-PN3 PEPPB-01HH-PS3 PEPPB-03GG-PN3 PEPPB-03GG-PS3 PEPPB-03HH-PN3 PEPPB-03HH-PS3 PEPPB-05GG-PN3 PEPPB-05GG-PS3 PEPPB-05HH-PN3 PEPPB-05HH-PS3 PEPPB-10GG-PN3 PEPPB-10GG-PS3 PEPPB-10HH-PN3 PEPPB-10HH-PS3 PEPPB-96GG-PN3 PEPPB-96GG-PS3 PEPPB-96HH-PN3 PEPPB-96HH-PS3 PEPPA-05HH-PN3 PEPPA-05HH-PS3 PEPPA-10GG-PN3 PEPPA-10GG-PS3 PEPPA-10HH-PN3 PEPPA-10HH-PS3 PEPPA-96GG-PN3 PEPPA-96GG-PS3 PEPPA-96HH-PN3 PEPPA-96HH-PS3 PEPPE-05HH-PN3 PEPPE-05HH-PS3 PEPPE-10GG-PN3 PEPPE-10GG-PS3 PEPPE-10HH-PN3 PEPPE-10HH-PS3 PEPPE-96GG-PN3 PEPPE-96GG-PS3 PEPPE-96HH-PN3 PEPPE-96HH-PS3 PEPPB-01GG-PS3-FB PEPPE-05GG-PN3-FB
76554-134PK 342.32 CAD
76554-134 76554-136 77296-340 77296-342 77296-344 77296-346 77296-348 77296-350 77296-352 77296-354 77296-356 77296-358 77296-420 77296-422 77296-424 77296-426 77296-428 77296-430 77296-432 77296-434 77296-436 77296-438 77296-380 77296-382 77296-384 77296-386 77296-388 77296-390 77296-392 77296-394 77296-396 77296-398 77296-400 77296-402 77296-404 77296-406 77296-408 77296-410 77296-412 77296-414 77296-416 77296-418 77296-360 77296-362 77296-364 77296-366 77296-368 77296-370 77296-372 77296-374 77296-376 77296-378 77296-440 77296-442 77296-444 77296-446 77296-448 77296-450 77296-452 77296-454 77296-456 77296-458 77437-870 77437-872
PROCLEAR Polypropylene DEMI Capsules
Filters Membrane Filter Capsules
PROCLEAR polypropylene filters are designed for a wide range of prefiltration duties within the production of pharmaceuticals. They are particularly suited for applications where chemical compatibility is an issue.
  • Graded density polypropylene media for high capacity
  • Extremely robust to withstand aggressive conditions
  • All polypropylene construction
  • MURUS and DEMICAPs can be gamma-irradiated and autoclaved
  • PFAS-free options available
The optimum pleat configuration and graded density polypropylene media used in PROCLEAR PP filters ensure the filters
More Product Information