BRAND® Mortars and Pestles
Supplier: Brandtech
110702 110703 110708 110709
77296-262EA 72.07 CAD
77296-262 77296-264 77296-266 77296-268
BRAND® Mortars and Pestles
Mortars and Pestles Mortars with Pestles
Mortars and pestles made from a durable melamine formaldehyde (MF) resin, ideal for grinding and crushing a variety of materials.
  • Autoclavable up to 121 °C
  • Exceptional resistance to boiling and scratching
  • Improved hardness, with a smooth surface texture
  • Mortars come equipped with a foot and spout for added convenience
  • Pestles are shaped for effortless gripping
Melamine formaldehyde resin, renowned for its thermal stability and remarkable resistance to various chemicals, makes BRAND® mortars and pestles ideal for grinding a diverse array of substancesMore Product Information