Industrial Label Printer with Brady Workstation Software Barcode Scanner Kit, i3300
Supplier: Brady Worldwide
176720 176704 176712
77282-208KT 3946.25 CAD
77282-208 77282-176 77282-192
Industrial Label Printer with Brady Workstation Software Barcode Scanner Kit, i3300
Printers Label Printers
Optimize your operations with a Brady printer and scanner system. Get a printer, scanner and software kit - advanced tools that make workflows more seamless and boost productivity.
  • Single color printing capability
  • Printer connectivity options: Ethernet; USB; USB 2.0; Wi-Fi
  • Auto cutter
  • PC connected
  • Scanner field of view: high density field: 30° horizontal by 20° vertical; wide field: 50° horizontal by 33.5° vertical
Get auto setup, auto calibration and fast changeovers with the i3300 label printer (300 dpi). Enjoy seamless, PrintSmart technology thatMore Product Information