Pegboards with ³/₁₆" Hole Size, White Polypropylene
Supplier: Triton Products
018
77269-190PK 44.76 CAD
77269-190
Pegboards with ³/₁₆" Hole Size, White Polypropylene
Furniture Backsplashes, Wallracks, Wallpanels
DuraBoard® is the most durable, weather and impact resistant pegboard available. It is 8 times stronger than conventional pegboard.
  • Hole diameter is ³/₁₆" with 1" O.C. spacing
  • DuraBoard will never fade, dry out, crack or peel over time so it maintains holding capability over many years of use
  • Offers a professional looking pegboard system that can be cleaned without harming the board
  • Board surface is ideal for creating silhouettes of tools to be stored and managed allowing for easy tool identification
  • Use Triton 70016 mounting spacer kit to mount DuraBoard
More Product Information