Anti-CD80 Rabbit Monoclonal Antibody [clone: ARC0384]

Supplier: ANTIBODIES.COM LLC
A309124-100
77232-638EA 781.15 CAD
77232-638 77232-640
Anti-CD80 Rabbit Monoclonal Antibody [clone: ARC0384]
Antibodies
Rabbit monoclonal [ARC0384] antibody to CD80 for WB with samples derived from Human.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2,000

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: ARC0384
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR