Anti-Estrogen Related Receptor alpha Rabbit Monoclonal Antibody [clone: ARC0919]

Supplier: ANTIBODIES.COM LLC
A307863-100
77227-664EA 781.15 CAD
77227-664 77227-666
Anti-Estrogen Related Receptor alpha Rabbit Monoclonal Antibody [clone: ARC0919]
Antibodies
Rabbit monoclonal [ARC0919] antibody to Estrogen Related Receptor alpha for WB with samples derived from Human and Mouse.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2,000

Type: Primary
Antigen: Estrogen Related Receptor alpha
Clonality: Monoclonal
Clone: ARC0919
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR