Anti-Galectin 1 Rabbit Monoclonal Antibody [Clone: ARC1110]

Supplier: ANTIBODIES.COM LLC
A306834-100
77223-600EA 781.15 CAD
77223-600 77223-602
Anti-Galectin 1 Rabbit Monoclonal Antibody [Clone: ARC1110]
Antibodies
Rabbit monoclonal [ARC1110] antibody to Galectin 1 for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC, ICC/IF
  • Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: Galectin 1
Clonality: Monoclonal
Clone: ARC1110
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR