Anti-Asparagine synthetase Rabbit Monoclonal Antibody [clone: ARC0517]

Supplier: ANTIBODIES.COM LLC
A305812-100
77219-564EA 481.9 CAD
77219-564 77219-562
Anti-Asparagine synthetase Rabbit Monoclonal Antibody [clone: ARC0517]
Antibodies
Rabbit monoclonal [ARC0517] antibody to Asparagine synthetase for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2,000

Type: Primary
Antigen: Asparagine synthetase
Clonality: Monoclonal
Clone: ARC0517
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR