Anti-TRK fused gene Rabbit Monoclonal Antibody [clone: ARC1882]

Supplier: ANTIBODIES.COM LLC
A305376-100
77217-834EA 781.15 CAD
77217-834 77217-836
Anti-TRK fused gene Rabbit Monoclonal Antibody [clone: ARC1882]
Antibodies
Rabbit monoclonal [ARC1882] antibody to TRK fused gene for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2,000

Type: Primary
Antigen: TRK fused gene
Clonality: Monoclonal
Clone: ARC1882
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR