Anti-Enterovirus 71 3D Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A8332-100
77212-040EA 555.69 CAD
77212-040 77215-426
Anti-Enterovirus 71 3D Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Enterovirus 71 3D for WB with samples derived from Enterovirus 71.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:100

Type: Primary
Antigen: Enterovirus 71 3D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Enterovirus 71
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR