Anti-Acetyl Coenzyme A carboxylase (phospho Ser79) Rabbit Monoclonal Antibody [clone: ARC56611]

Supplier: ANTIBODIES.COM LLC
A309566-100
77210-902EA 901.86 CAD
77210-902 77211-248
Anti-Acetyl Coenzyme A carboxylase (phospho Ser79) Rabbit Monoclonal Antibody [clone: ARC56611]
Antibodies
Rabbit monoclonal [ARC56611] antibody to Acetyl Coenzyme A carboxylase (phospho Ser79) for WB with samples derived from Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:2,000 to 1:8,000

Type: Primary
Antigen: Acetyl Coenzyme A carboxylase
Clonality: Monoclonal
Clone: ARC56611
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR